Learn More
Accuflow Systems 50ML BAG 2 PORT W12" TUB 20/PK
Catalog No. NC1288664
Encompass_Preferred
Standard 2 port 50ml bag with 12” 18”x 1/4" Cflex tubing on each port with male and female Luer Locks. pk/20
Catalog No. NC1288664 Supplier Accuflow Systems Supplier No. BP0050MS
Standard 2 port 50ml bag with 12” 18”x 1/4" Cflex tubing on each port with male and female Luer Locks. pk/20