Learn More
US Plastic Corporation 50ML BEAKERS PP DISP 100/PK
SDP
Catalog No. NC0968025
Encompass
50ml Graduated Beakers, Polypropylene, Dripless lip & flat base, Molded-in graduations in ml & oz., Translucent, autoclavable, Maximum temperature: 250a Deg. F. 100 per pack.
Catalog No. NC0968025 Supplier US Plastic Corporation Supplier No. 76053
50ml Graduated Beakers, Polypropylene, Dripless lip & flat base, Molded-in graduations in ml & oz., Translucent, autoclavable, Maximum temperature: 250a Deg. F. 100 per pack.