Learn More
CELLTREAT Scientific Products 50mL Self-Standing Bio-Reaction Tube - Bag, Sterile
SDP
Catalog No. 50202015
Encompass_Preferred
CELLTREAT Scientific Products 50mL Bio-reaction Tube Self Standing. Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture. Ideal for biological reaction applications and free gas exchange. Aseptic conditions maintained through 0.22?m hydrophobic vented membrane. Useful in the screening and optimization processes of suspension cells. Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously. Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization. Easy to read black graduations and a large white marking area. RNase/DNase free and non-pyrogenic. Max force (RCF) g 16000. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 100/case.
Catalog No. 50-202-015 Supplier CELLTREAT Scientific Products Supplier No. 229479
CELLTREAT Scientific Products 50mL Bio-reaction Tube Self Standing. Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture. Ideal for biological reaction applications and free gas exchange. Aseptic conditions maintained through 0.22?m hydrophobic vented membrane. Useful in the screening and optimization processes of suspension cells. Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously. Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization. Easy to read black graduations and a large white marking area. RNase/DNase free and non-pyrogenic. Max force (RCF) g 16000. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 100/case.