Learn More
Quality Biological Inc 50 mL Centrifuge Tube and Cap - Non-sterile (Tubes and Caps Packed Separately), 500 Tubes/CS and 500 Caps/CS
SDP
Catalog No. 50147450
Encompass_Preferred
50 mL Centrifuge Tube and Cap - Non-Ssterile (Tubes and Caps Packed Separately)
Packaged 500 Tubes/Bag and 500 Caps/Bag, 500/CS
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%, 2.5 mL graduation increments
• Large white marking area provides excellent contrast for identification
• Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
Catalog No. 50-147-450 Supplier Quality Biological Inc Supplier No. P886229456
50 mL Centrifuge Tube and Cap - Non-Ssterile (Tubes and Caps Packed Separately)
Packaged 500 Tubes/Bag and 500 Caps/Bag, 500/CS
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%, 2.5 mL graduation increments
• Large white marking area provides excellent contrast for identification
• Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification