Learn More
Quality Biological Inc 50 mL Centrifuge Tube with Conical Bottom - Non-sterile, Paperboard Rack, 25/Rack, 500/CS
SDP
Catalog No. 50147449
Encompass_Preferred
50 mL Centrifuge Tube with Conical Bottom - Non-Sterile
Packaged 25/Paperboard Rack, 500/CS
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
• Large white marking area provides excellent contrast for identification
• Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
• Convenient zip-closure bag is a practical alternative to folding and taping opened bags
• Paperboard rack is a recyclable, fold-flat alternative to bulky foam racks
• Non-Sterile option
Catalog No. 50-147-449 Supplier Quality Biological Inc Supplier No. P886229455
50 mL Centrifuge Tube with Conical Bottom - Non-Sterile
Packaged 25/Paperboard Rack, 500/CS
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
• Large white marking area provides excellent contrast for identification
• Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
• Convenient zip-closure bag is a practical alternative to folding and taping opened bags
• Paperboard rack is a recyclable, fold-flat alternative to bulky foam racks
• Non-Sterile option