Learn More
Phenomenex Inc 50mL Centrifugal Tube
Catalog No. 502398731 Shop All Phenomenex Products
Encompass_Preferred
50mL Centrifugal Tube 1g MgSO4 (Anhydrous), 0.5g NaAc
Catalog No. 50-239-8731 Supplier Phenomenex Inc Supplier No. MSMG5064
50mL Centrifugal Tube 1g MgSO4 (Anhydrous), 0.5g NaAc