Learn More
CELLTREAT Scientific Products CAP ONLY, 50mL Centrifuge Tube Cap - Bag, Non-sterile
SDP
Catalog No. 50202019
Encompass_Preferred
CELLTREAT Scientific Products 50mL Centrifuge Tube CAP ONLY. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Cap Only for 15mL Centrifuge Tube. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/bag, 500/case.
Catalog No. 50-202-019 Supplier CELLTREAT Scientific Products Supplier No. 229453
CELLTREAT Scientific Products 50mL Centrifuge Tube CAP ONLY. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Cap Only for 15mL Centrifuge Tube. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/bag, 500/case.