Learn More
Bellco Glass, Inc. 50mL Centrifuge Tube
SDP
Catalog No. 502329615
Encompass_Preferred
Graduations are printed in bold easytoread black ink and are accurate to plus/2. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene HDPE caps feature integrated leakproof seal. Tubes include graduations molded into the conical bottom ideal for pellet viewing and small volume quantification. Convenient zipclosure bag is a practical alternative to folding and taping opened bags. Assorted color cap tubes allow easily color coding of samples for quick identification customization and increased productivity. Users report coding each color to a day of the week a member of the lab or a specific procedure. Each case of assorted color tubes 230411A and 230421A comes with a free plastic rack and 4 bags of each individual color red yellow green blue purple Specifications Plastic rack is manufactured from polypropylene and is autoclavable reusable and recyclable <
Catalog No. 50-232-9615 Supplier Bellco Glass, Inc. Supplier No. 230421R00
Graduations are printed in bold easytoread black ink and are accurate to plus/2. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene HDPE caps feature integrated leakproof seal. Tubes include graduations molded into the conical bottom ideal for pellet viewing and small volume quantification. Convenient zipclosure bag is a practical alternative to folding and taping opened bags. Assorted color cap tubes allow easily color coding of samples for quick identification customization and increased productivity. Users report coding each color to a day of the week a member of the lab or a specific procedure. Each case of assorted color tubes 230411A and 230421A comes with a free plastic rack and 4 bags of each individual color red yellow green blue purple Specifications Plastic rack is manufactured from polypropylene and is autoclavable reusable and recyclable <