Learn More
Quality Biological Inc 50 mL Centrifuge Tube only, Conical Bottom - Non-sterile, Bag - 500/CS
SDP
Catalog No. 50147448
Encompass_Preferred
50 mL Centrifuge TUBE ONLY, Conical Bottom - Non-Sterile
Packaged 500/BAG, 500/CS
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
• Large white marking area provides excellent contrast for identification
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
• Tube Only
Catalog No. 50-147-448 Supplier Quality Biological Inc Supplier No. P886229454
50 mL Centrifuge TUBE ONLY, Conical Bottom - Non-Sterile
Packaged 500/BAG, 500/CS
• Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
• Large white marking area provides excellent contrast for identification
• Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
• Tube Only