Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube - Bag, Sterile
SDP
Catalog No. NC2089370
Encompass_Preferred
Manufactured and packaged using an improved process, while maintaining standards. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Large white marking area provides excellent contrast for identification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags.
This item is not returnable. View return policy
Catalog No. NC2089370 Supplier CELLTREAT Scientific Products Supplier No. 229435
Manufactured and packaged using an improved process, while maintaining standards. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Large white marking area provides excellent contrast for identification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags.