Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube - Black, Bag, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229433
Encompass_Preferred
Centrifuge Tube, 50mL, conical bottom. CELLTREAT Scientific Products. Opaque black polypropylene protects light sensitive compounds, 12,000 (RCF)g. Large white marking area. Sterile (gamma irradiated). Bulk Pack, 25/Re-sealable bag, 100/CS.
Catalog No. 50-550-489