Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube, Flip Top Cap - Paperboard Rack, Sterile
SDP
Catalog No. 50202005
Encompass_Preferred
CELLTREAT Scientific Products 50mL Centrifuge Tube with Flip-top Cap in Paperboard Rack. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Ergonomic flip top cap is designed for single handed operation. Paperboard rack is a recyclable, fold-flat alternative to bulky foam racks. RNase/DNase free and non-pyrogenic. Max force (RCF) g 9400. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/paperboard rack, 500/case.
Catalog No. 50-202-005 Supplier CELLTREAT Scientific Products Supplier No. 229426
CELLTREAT Scientific Products 50mL Centrifuge Tube with Flip-top Cap in Paperboard Rack. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Ergonomic flip top cap is designed for single handed operation. Paperboard rack is a recyclable, fold-flat alternative to bulky foam racks. RNase/DNase free and non-pyrogenic. Max force (RCF) g 9400. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/paperboard rack, 500/case.