Learn More
Chemglass Life Sciences Centrifuge Tube, 50mL, Graduated, Polypropylene, Sterile, Bulk Pack
SDP
Catalog No. 501897639
Encompass_Preferred
• Printed graduations (with marking area)• Polypropylene tube with polyethylene cap• Flat markable cap with leak-proof seal• Easy, one-handed, cap operation• Sterilized by E-Beam• Non-pyrogenic, DNase- and RNase-free• Packed in bags of 25• Max. RCF (xg) 12,000
Catalog No. 50-189-7639 Supplier Chemglass Life Sciences Supplier No. CGN4303050
• Printed graduations (with marking area)• Polypropylene tube with polyethylene cap• Flat markable cap with leak-proof seal• Easy, one-handed, cap operation• Sterilized by E-Beam• Non-pyrogenic, DNase- and RNase-free• Packed in bags of 25• Max. RCF (xg) 12,000