Learn More
CELLTREAT Scientific Products TUBE ONLY, 50mL Centrifuge Tube - Bag, Non-sterile
SDP
Catalog No. 50202020
Encompass_Preferred
CELLTREAT Scientific Products 50mL Centrifuge Tube, TUBE ONLY. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Tube Only. RNase/DNase free and non-pyrogenic. Max force (RCF) g 16000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/bag, 500/case.
Catalog No. 50-202-020 Supplier CELLTREAT Scientific Products Supplier No. 229454
CELLTREAT Scientific Products 50mL Centrifuge Tube, TUBE ONLY. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Tube Only. RNase/DNase free and non-pyrogenic. Max force (RCF) g 16000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/bag, 500/case.