Learn More
CELLTREAT Scientific Products PLASTIC RACK ONLY, 50mL Centrifuge Tube, Reusable, Non-sterile
SDP
Catalog No. 50607517
Encompass_Preferred
CELLTREAT Scientific Products 50mL Centrifuge Tube PLASTIC RACK ONLY. Plastic rack is manufactured from polypropylene and is autoclvable, reusable, and recyclable. Chemically resistant to drips. Design of bottom prevents racks from sticking/freezing to surfaces. Widely spaced design catches drips and allows tubes to be retrieved more easily. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 5/case.
Catalog No. 50-607-517 Supplier CELLTREAT Scientific Products Supplier No. 229429
CELLTREAT Scientific Products 50mL Centrifuge Tube PLASTIC RACK ONLY. Plastic rack is manufactured from polypropylene and is autoclvable, reusable, and recyclable. Chemically resistant to drips. Design of bottom prevents racks from sticking/freezing to surfaces. Widely spaced design catches drips and allows tubes to be retrieved more easily. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 5/case.