Learn More
Alkali Scientific CellPro™ 50mL Centrifuge Tube Rack, Fits 25 x 50mL, Sterile, 1/pk
SDP
Catalog No. 502389535
Encompass_Preferred
CellPro™ 50mL Centrifuge Tube Rack is designed to hold 25 50mL tubes securely. Made from durable, sterile, RNase, DNase and pyrogen-free polypropylene, it ensures stability during transport and storage. With precise slot sizing, organizing and storing samples is effortless.
Catalog No. 50-238-9535 Supplier Alkali Scientific Supplier No. CN5610
CellPro™ 50mL Centrifuge Tube Rack is designed to hold 25 50mL tubes securely. Made from durable, sterile, RNase, DNase and pyrogen-free polypropylene, it ensures stability during transport and storage. With precise slot sizing, organizing and storing samples is effortless.