Learn More
CEM Corporation 50 mL Polypropylene Centrifuge Tubes (pack of 25)
SDP
Catalog No. 501877683
Encompass
The 50 mL Polypropylene Centrifuge Tubes (pack of 25) Used to: hold the amino acids in solution on the reagent manifolds of the Liberty Blue, collect peptide after TFA cleavage on the Razor Parallel Cleavage System and the Discover Bio wash station, collect extracted samples on the EDGE.
Catalog No. 50-187-7683 Supplier CEM Corporation Supplier No. 330090
The 50 mL Polypropylene Centrifuge Tubes (pack of 25) Used to: hold the amino acids in solution on the reagent manifolds of the Liberty Blue, collect peptide after TFA cleavage on the Razor Parallel Cleavage System and the Discover Bio wash station, collect extracted samples on the EDGE.