Learn More
Azzota Corp 50Ml Centrifuge Tubes, Made Of High-Clarity Polypropylene. Allowable Temperature From -80Deg.C To 120Deg.C. Can Sustain 12,000G, Conical Bottom, 25 Tubes And Caps Per Bag.
Catalog No. 507532880
Encompass_Preferred
50ml Centrifuge Tubes, Made of high-clarity polypropylene. Allowable temperature from -80deg.C to 120deg.C. Can sustain 12,000g, Conical Bottom, 25 tubes and caps per bag.
Catalog No. 50-753-2880 Supplier Azzota Corp Supplier No. CT50
50ml Centrifuge Tubes, Made of high-clarity polypropylene. Allowable temperature from -80deg.C to 120deg.C. Can sustain 12,000g, Conical Bottom, 25 tubes and caps per bag.