Learn More
Genesee Scientific Corporation 50 mL Conical Centrifuge Tubes, Sterile, Gamma Irradiated, Bulk 500 Units
SDP
Catalog No. NC1259986
Encompass_Preferred
50 mL Conical Centrifuge Tubes from Genesee Scientific are ideal for diverse scientific applications. Made from medical-grade virgin polypropylene, they are chemically resistant and ensure sample integrity. With leak-proof screw caps and clear graduations, these sterile tubes can endure maximum RCF of 15,000 x g, suitable for high-speed centrifugation and cryogenic storage down to -80°C. Packaged in bulk, they provide exceptional value and reliability for critical research needs.
Catalog No. NC1259986 Supplier Genesee Scientific Corporation Supplier No. 28108
50 mL Conical Centrifuge Tubes from Genesee Scientific are ideal for diverse scientific applications. Made from medical-grade virgin polypropylene, they are chemically resistant and ensure sample integrity. With leak-proof screw caps and clear graduations, these sterile tubes can endure maximum RCF of 15,000 x g, suitable for high-speed centrifugation and cryogenic storage down to -80°C. Packaged in bulk, they provide exceptional value and reliability for critical research needs.