Learn More
Alkali Scientific ASI™ 50mL Centrifuge Tubes, Conical, Foam Rack, Sterile, Flat Blue Screw Cap, PP (500/Cs)
SDP
Catalog No. 503026524
Encompass_Preferred
Sterile High centrifugal speeds (9,500 x g) Packaged in bags of 25 / 500 per case Secure seal under cap to prevent leakage. Easy-to-read graduations Can be written on without smudging Flat caps with green color Certified RNase/DNase, pyrogen free
Catalog No. 50-302-6524 Supplier Alkali Scientific Supplier No. C5602
Sterile High centrifugal speeds (9,500 x g) Packaged in bags of 25 / 500 per case Secure seal under cap to prevent leakage. Easy-to-read graduations Can be written on without smudging Flat caps with green color Certified RNase/DNase, pyrogen free