Learn More
Beckman Coulter 30.0 MM Diameter Polypropylene Conical Tube Adapter, 50 mL, 6/pk
Catalog No. NC1324406
Encompass
For use in benchtop and microcentrifuge rotors: FX6100 F0685 Color: beige. Quantity of 6.
Catalog No. NC1324406 Supplier Beckman Coulter Supplier No. 392268
For use in benchtop and microcentrifuge rotors: FX6100 F0685 Color: beige. Quantity of 6.