Learn More
Waters Corp Centrifuge Tube Domino, 50 mL Conical, 8 Tube Capacity
Catalog No. NC3597557 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Centrifuge Tube Domino is a machine-readable consumable holder designed to interface various consumable types with automated pipetting robots. It holds up to 8 standard, non self-standing 50 mL conical tubes, enabling seamless automation of laboratory workflows.
  • Holds up to 8 standard, non self-standing 50 mL tubes.
  • Designed for use with the Andrew+ pipetting robot.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
Catalog No. NC3597557 Supplier Waters Corp Supplier No. 186009614
The Centrifuge Tube Domino is a machine-readable consumable holder designed to interface various consumable types with automated pipetting robots. It holds up to 8 standard, non self-standing 50 mL conical tubes, enabling seamless automation of laboratory workflows.
  • Holds up to 8 standard, non self-standing 50 mL tubes.
  • Designed for use with the Andrew+ pipetting robot.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.