Learn More
S BIO ProteoSave™ 50mL Conicaltube
Catalog No. NC2054213
Encompass
ProteoSave™ products feature surfaces coated with hydrophilic polymer to reduce nonspecific adsorption of biological samples such as peptides and proteins. Widely used in biological research and biomarker discovery applications, these tubes provide multiple advantages: they reduce protein, peptide, and cell adsorption; resist organic solvents, detergents, and heat treatment (boiling at 100°C for 10 minutes); and come sterilized for suitability in preserving cell products, additives, and freeze-thaw cycles.
Catalog No. NC2054213 Supplier S BIO Supplier No. MS52550
ProteoSave™ products feature surfaces coated with hydrophilic polymer to reduce nonspecific adsorption of biological samples such as peptides and proteins. Widely used in biological research and biomarker discovery applications, these tubes provide multiple advantages: they reduce protein, peptide, and cell adsorption; resist organic solvents, detergents, and heat treatment (boiling at 100°C for 10 minutes); and come sterilized for suitability in preserving cell products, additives, and freeze-thaw cycles.