Learn More
Charter Medical 50ML CRYO FREEZE BAG 25/CS
Catalog No. NC1122965
Encompass
Manufactured by Incell Corporation and supplied by Charter Medical:50mL max fill volume, (1) FP-FLEX lines (8 L ; 1/8ID x 1/4 OD) sealed; 1 EVA line with 2 PVC leads (4 L ; 0.118 ID x 0.160 OD) and female luers. Volume: 10 30mL Tubing and containers tested and validated to -196CFP-FLEX tubing compatible with C-FlexClosed-system aseptic transfer via tube-to-tube connection
Catalog No. NC1122965 Supplier Charter Medical Supplier No. FPFLEX50B
Manufactured by Incell Corporation and supplied by Charter Medical:50mL max fill volume, (1) FP-FLEX lines (8 L ; 1/8ID x 1/4 OD) sealed; 1 EVA line with 2 PVC leads (4 L ; 0.118 ID x 0.160 OD) and female luers. Volume: 10 30mL Tubing and containers tested and validated to -196CFP-FLEX tubing compatible with C-FlexClosed-system aseptic transfer via tube-to-tube connection