Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube, Blue Cap - Bag, Sterile, 500/CS
SDP
Catalog No. 501535102
Encompass_Preferred
CELLTREAT Scientific Products 50mL Centrifuge Tube, Blue Cap. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Polypropylene tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Assorted color cap tubes allow easily color coding of samples for quick identification, customization, and increased productivity. Users report coding each color to a day of the week, a member of the lab, or a specific procedure. Max RCF: 16,000g. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/resealable bag, 500/case.
Catalog No. 50-153-5102 Supplier CELLTREAT Scientific Products Supplier No. 230421B
CELLTREAT Scientific Products 50mL Centrifuge Tube, Blue Cap. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Polypropylene tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Assorted color cap tubes allow easily color coding of samples for quick identification, customization, and increased productivity. Users report coding each color to a day of the week, a member of the lab, or a specific procedure. Max RCF: 16,000g. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/resealable bag, 500/case.