Learn More
S2 Media 50mL Double Strength Tryptic Soy Broth (2x TSB), 120mL Flip-Cap Tube, 1/each
Catalog No. NC3900344
Encompass_Preferred
This item has a minimum order quantity of 10 per supplier requirements
Double-strength Tryptic Soy Broth (TSB) is a general nutrient and enrichment broth. It is used in qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms and is also recommended for sterility testing. TSB is recommended for use as the inoculum broth for disc diffusion and agar dilution antimicrobial susceptibility testing by the CLSI. Reference the appropriate standard method for additional details on test instructions. • Product meets AOAC, BAM, CLSI, EPA, ISO, NF, SM-Dairy, SMWW, USDA, and Harmonized USP/EP/JP reference methods and performance criteria.• TSB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Soybean-Casein Digest (SCD) Medium.• Double Strength (2x) Formulation.
Catalog No. NC3900344 Supplier S2 Media Supplier No. 5522
This item has a minimum order quantity of 10 per supplier requirements
Double-strength Tryptic Soy Broth (TSB) is a general nutrient and enrichment broth. It is used in qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms and is also recommended for sterility testing. TSB is recommended for use as the inoculum broth for disc diffusion and agar dilution antimicrobial susceptibility testing by the CLSI. Reference the appropriate standard method for additional details on test instructions. • Product meets AOAC, BAM, CLSI, EPA, ISO, NF, SM-Dairy, SMWW, USDA, and Harmonized USP/EP/JP reference methods and performance criteria.• TSB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Soybean-Casein Digest (SCD) Medium.• Double Strength (2x) Formulation.