Learn More
Stellar Scientific iTubes 50mL Conical Tubes, Rnase/DNase Free, STERILE Foam Racked, 500/cs
SDP
Catalog No. NC1637810
Encompass
Made or premium polypropylene with uniformly thick walls for thickness and clarity. Max RCF of 12,000xg (for 15mL tubes) and 15,000xg (for 50mL tubes). Safe to store at temperatures down to -80°C. Sterilized with ethaline oxide. Tubes and caps are certified for stem cell research. Frosted writing area, and lid is super-flat and frosted for easy writing. RNase, DNase, Pyrogen free.
Catalog No. NC1637810 Supplier Stellar Scientific Supplier No. T50600
Made or premium polypropylene with uniformly thick walls for thickness and clarity. Max RCF of 12,000xg (for 15mL tubes) and 15,000xg (for 50mL tubes). Safe to store at temperatures down to -80°C. Sterilized with ethaline oxide. Tubes and caps are certified for stem cell research. Frosted writing area, and lid is super-flat and frosted for easy writing. RNase, DNase, Pyrogen free.