Learn More
Alkali Scientific 500mL Erlenmeyer Flask with Plug Seal Cap and Flat Bottom, PETG Plastic, Sterile, 12/cs
SDP
Catalog No. 502336231
Encompass_Preferred
Our Erlenmeyer Flasks are made from PETG with an HDPE cap and are manufactured in air filtered clean rooms. These Erlenmeyer Flasks are ideal for shaker culture applications. Erlenmeyer Flasks are sterilized by E-Beam and are non-pyrogenic, DNase and RNase free. They are non-toxic and environmentally friendly. They come packaged individually in a sterile zip sealable bag.
Catalog No. 50-233-6231 Supplier Alkali Scientific Supplier No. EFN704
Our Erlenmeyer Flasks are made from PETG with an HDPE cap and are manufactured in air filtered clean rooms. These Erlenmeyer Flasks are ideal for shaker culture applications. Erlenmeyer Flasks are sterilized by E-Beam and are non-pyrogenic, DNase and RNase free. They are non-toxic and environmentally friendly. They come packaged individually in a sterile zip sealable bag.