Learn More
Chemglass Life Sciences 50mL Erlenmeyer Flask, 24-410 GPI Screw Thread
SDP
Catalog No. CG154301
Encompass_Preferred
50mL Erlenmeyer Flask, 24-410 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. CG154301 Supplier Chemglass Life Sciences Supplier No. CG154301
50mL Erlenmeyer Flask, 24-410 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.