Learn More
Chemglass Life Sciences 50mL Glassblowers RBF Blank
SDP
Catalog No. 501531796
Encompass_Preferred
Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-153-1796 Supplier Chemglass Life Sciences Supplier No. CG61802
Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.