Learn More
Bellco Glass, Inc. 50mL Flask Tray, 25x25, 16 Pos
SDP
Catalog No. 501946135
Encompass_Preferred
Pre-mounted stainless steel clamps for Bellco Erlenmeyer flask. Flask Tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 16 Dimensions: 25x25mm Size: 50ml
Catalog No. 50-194-6135 Supplier Bellco Glass, Inc. Supplier No. 774405016
Pre-mounted stainless steel clamps for Bellco Erlenmeyer flask. Flask Tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 16 Dimensions: 25x25mm Size: 50ml