Learn More
Hach Company 50mL Flask-Volumet-Polypro
Supplier:  Hach Company 1406041
Encompass
Polypropylene flask and screw cap Durable with excellent chemical resistance with the same accuracy as glass Permanent color graduation line with marking spot Calibrated to contain/deliver at 20 C
Catalog No. 50-002-57105