Learn More
CELLTREAT Scientific Products 50mL Graduated Cylinder, Polymethylpentene, Non-sterile, Case Quantity 6
SDP
Catalog No. 5000670049
Encompass_Preferred
Graduated Cylinders are designed with ring-molded, incrementalgraduations to ensure excellent readability and accuracy
Premium polymethylpentene material is a transparent,lightweight, and shatterproof alternative to glass measuringcylinders, with strong chemical resistance and a maximumtemperature tolerance of 135 ̊C
Pour spout and rolled lip design prevent drips and splashes
Cylinders feature a wide hexagonal base for increased stability
Catalog No. 50-006-70049 Supplier CELLTREAT Scientific Products Supplier No. 230533
Graduated Cylinders are designed with ring-molded, incrementalgraduations to ensure excellent readability and accuracy
Premium polymethylpentene material is a transparent,lightweight, and shatterproof alternative to glass measuringcylinders, with strong chemical resistance and a maximumtemperature tolerance of 135 ̊C
Pour spout and rolled lip design prevent drips and splashes
Cylinders feature a wide hexagonal base for increased stability