Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0766908
Encompass_Preferred
Hemispherical fabric heating mantle for 50ml single or multi neck flasks to 400 degrees C maximum internal operating temperatrure, 60W, 115V, CSA.
Catalog No. NC0766908 Supplier Glas Col LLC Supplier No. 100A O394
Hemispherical fabric heating mantle for 50ml single or multi neck flasks to 400 degrees C maximum internal operating temperatrure, 60W, 115V, CSA.