Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO Glass Reactor EM 50mL, 5 ports

Catalog No. NC2054062 Shop All METTLER TOLEDO Products
Encompass

Glass Reactor EasyMax 50 mL, 5 ports

Catalog No. NC2054062 Supplier METTLER TOLEDO Supplier No. 51161792
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glass Reactor EasyMax 50 mL, 5 ports

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.