Learn More
Chemglass Life Sciences 50mL Peptide Synthesis Vessel, Solid Phase, 2mm T-Bore PTFE Stpk, Medium Frit, GL 25 Thread
SDP
Catalog No. CG186403
Encompass_Preferred
50mL Peptide Synthesis Vessel, Solid Phase, 2mm T-Bore PTFE Stpk, Medium Frit, GL 25 Thread. Solid phase peptide synthesis vessel having a medium porosity, fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower stopcock is a “T”-Bore 2mm PTFE to allow nitrogen to be used to upwardly agitate the resin beads while shaking. The O.D. of both lower stems is 8mm. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186403 Supplier Chemglass Life Sciences Supplier No. CG186403
50mL Peptide Synthesis Vessel, Solid Phase, 2mm T-Bore PTFE Stpk, Medium Frit, GL 25 Thread. Solid phase peptide synthesis vessel having a medium porosity, fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower stopcock is a “T”-Bore 2mm PTFE to allow nitrogen to be used to upwardly agitate the resin beads while shaking. The O.D. of both lower stems is 8mm. Vessels are available in sizes other than those listed. Please call for details.