Learn More
Chemglass Life Sciences 50mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Medium Frit, GL 25 Thread
SDP
Catalog No. CG186603
Encompass_Preferred
50mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Medium Frit, GL 25 Thread. Similar to CG-1864, but with the addition of a lower standard taper inner joint to allow collection into a vessel having a 24/40 outer joint. MEDIUM porosity fritted glass resin support. A serrated hose connection is sealed on the inner joint for connection to a vacuum source. The “T”-Bore stopcock has an 8mm O.D. sidearm for connection to nitrogen, while the serrated hose connection has an O.D. of 8mm at the middle ring.
Catalog No. CG186603 Supplier Chemglass Life Sciences Supplier No. CG186603
50mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Medium Frit, GL 25 Thread. Similar to CG-1864, but with the addition of a lower standard taper inner joint to allow collection into a vessel having a 24/40 outer joint. MEDIUM porosity fritted glass resin support. A serrated hose connection is sealed on the inner joint for connection to a vacuum source. The “T”-Bore stopcock has an 8mm O.D. sidearm for connection to nitrogen, while the serrated hose connection has an O.D. of 8mm at the middle ring.