Learn More
Ace Glass, Inc. 50ML PESTLE ONLY 50ML PESTLE ONLY
SDP
Catalog No. 50974573
Encompass_Preferred
50mL glass Pestle with 10mm rod, for use with 50mL tapered tube (50974572) as a tissue grinder. The rod can be used with a 509558293 Flex-Grip Chuck.
This item is not returnable. View return policy
Catalog No. 50-974-573 Supplier Ace Glass, Inc. Supplier No. 832530
50mL glass Pestle with 10mm rod, for use with 50mL tapered tube (50974572) as a tissue grinder. The rod can be used with a 509558293 Flex-Grip Chuck.