Learn More
Savillex Corporation 50 mL PFA Dropper Bottle
SDP
Catalog No. NC1612143
Encompass
All PFA construction – the only PFA dropper bottle available. Microbore PFA tip for fine droplet control. Average droplet size 15 uL. Narrow droplet range: 13-18 uL. PFA cover prevents contamination and minimizes evaporation. Perfect for delivering isotope spike solutions and many other uses.
Catalog No. NC1612143 Supplier Savillex Corporation Supplier No. 700550
All PFA construction – the only PFA dropper bottle available. Microbore PFA tip for fine droplet control. Average droplet size 15 uL. Narrow droplet range: 13-18 uL. PFA cover prevents contamination and minimizes evaporation. Perfect for delivering isotope spike solutions and many other uses.