Learn More
Bellco Glass, Inc. 50mL Pipet Individually
SDP
Catalog No. 502329467
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Peeltoopen plastic wrappers are the ideal option for eliminating particulate proven benefit over traditional paper wrapper Wrappers reduce static cling versus other brands of plastic wraps Can be wiped with alcohol without smearing ink or penetrating paper 100 Individually Wrapped Case Quantity 800
Catalog No. 50-232-9467 Supplier Bellco Glass, Inc. Supplier No. 229230A00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Peeltoopen plastic wrappers are the ideal option for eliminating particulate proven benefit over traditional paper wrapper Wrappers reduce static cling versus other brands of plastic wraps Can be wiped with alcohol without smearing ink or penetrating paper 100 Individually Wrapped Case Quantity 800