Learn More
Bellco Glass, Inc. 50mL Pipet, Individually
SDP
Supplier:  Bellco Glass, Inc. 229230B00
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description30 Individually Wrapped/BagCase Quantity90SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorPurplePrimary MaterialPolystyreneMaximum Volume50mLGraduatedYesGraduation Increments (Subdivisions)5/10mLRNase/DNase FreeYes
Catalog No. 50-232-9958