Learn More
CELLTREAT Scientific Products 50mL Polystyrene Centrifuge Tube - Plastic Rack, Sterile
SDP
Catalog No. 5000144320
Encompass_Preferred
Polystyrene is optically clear for easy viewing of sample or colorimetric testingTubes feature molded graduations Easy to write on flat surface polyethylene (HDPE) capsPlastic rack is manufactured from polypropylene and is autoclavable, reusable, and recyclableDesign of bottom prevents racks from sticking/freezing to surfacesWidely spaced design catches drips and allows tubes to be retrieved more easily
Catalog No. 50-001-44320 Supplier CELLTREAT Scientific Products Supplier No. 229490
Polystyrene is optically clear for easy viewing of sample or colorimetric testingTubes feature molded graduations Easy to write on flat surface polyethylene (HDPE) capsPlastic rack is manufactured from polypropylene and is autoclavable, reusable, and recyclableDesign of bottom prevents racks from sticking/freezing to surfacesWidely spaced design catches drips and allows tubes to be retrieved more easily