Learn More
Chemglass Life Sciences 50mL Powder Addition Funnel, 14/20 Joint Size, Auger A
SDP
Catalog No. CG172305
Encompass_Preferred
50mL Powder Addition Funnel, 14/20 Joint Size, Auger A. Facilitates the delivery of free-flowing, granular solids into reactions. Pressure equalizing arm is equipped with a vented stopper for air-free operations and also permits access into the delivery stem for easy clean-out. One piece, easily removed grooved PTFE auger delivers powders at a uniform rate. Funnel has a lower inner joint and top outer, both being the size listed.
Catalog No. CG172305 Supplier Chemglass Life Sciences Supplier No. CG172305
50mL Powder Addition Funnel, 14/20 Joint Size, Auger A. Facilitates the delivery of free-flowing, granular solids into reactions. Pressure equalizing arm is equipped with a vented stopper for air-free operations and also permits access into the delivery stem for easy clean-out. One piece, easily removed grooved PTFE auger delivers powders at a uniform rate. Funnel has a lower inner joint and top outer, both being the size listed.