Learn More
Ace Glass, Inc. 50mL powder dispensing funnel vertical type
SDP
Catalog No. 50979222
Encompass_Preferred
50mL vertical, compact, screw-feed powder dispensing funnel. Features a flexible PTFE screw thread, a #15 Ace-Thred at the top, and a large, 1-1/4 knob. Top side port is 14/20, bottom joint is 24/40. 9-1/2 overall height.
Catalog No. 50-979-222 Supplier Ace Glass, Inc. Supplier No. 723320
50mL vertical, compact, screw-feed powder dispensing funnel. Features a flexible PTFE screw thread, a #15 Ace-Thred at the top, and a large, 1-1/4 knob. Top side port is 14/20, bottom joint is 24/40. 9-1/2 overall height.