Learn More
Alkali Scientific 50mL Centrifuge Tubes, PP (29 X 115mm), Flat Screw Cap, Individually Wrapped, Sterile
SDP
Catalog No. 502388749
Encompass_Preferred
50mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free
Catalog No. 50-238-8749 Supplier Alkali Scientific Supplier No. C5650W
50mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free