Learn More
CELLTREAT Scientific Products 50mL Graduated Beaker, Polypropylene, Non-sterile
SDP
Catalog No. 501656704
Encompass_Preferred
Molded pouring spouts and rolled lip for dripless work environment. Angled walls for easier handling and sample retrieval. Nesting design for easy stacking and storage. Smaller case quantities are easier to store and are ideal for small scale users. Polypropylene material for resistance to a variety of strong acids, bases and solvents. Every case is printed with the lot number for quality traceability. Packed non-sterile, 100/case.
Catalog No. 50-165-6704 Supplier CELLTREAT Scientific Products Supplier No. 230512
Molded pouring spouts and rolled lip for dripless work environment. Angled walls for easier handling and sample retrieval. Nesting design for easy stacking and storage. Smaller case quantities are easier to store and are ideal for small scale users. Polypropylene material for resistance to a variety of strong acids, bases and solvents. Every case is printed with the lot number for quality traceability. Packed non-sterile, 100/case.