Learn More
Labcon North America ProtectR Dry Ice Storage Tubes 50mL, 100/case
Catalog No. NC1704300
Encompass_Preferred
ProtectR™ tubes are the ideal vessel for primary and secondary sample containment. Samples are shipped on dry ice in a variety of volumes. When shipped in standard, small volume tubes, samples are at risk of acidifcation. ProtectR™ tubes can be used as a secondary shipping tube for multiple small volume tubes. With their airtight, leak-resistant secure seal, ProtectR™ tubes provide a protective barrier against CO2 exposure, safeguarding the sample(s) inside from costly damage. Only ProtectR™ tubes ensure consistent sample integrity and protein activity across experiments. Even if samples are delayed during shipping, they will not become compromised, saving time and money.
Catalog No. NC1704300 Supplier Labcon North America Supplier No. 38803250009
ProtectR™ tubes are the ideal vessel for primary and secondary sample containment. Samples are shipped on dry ice in a variety of volumes. When shipped in standard, small volume tubes, samples are at risk of acidifcation. ProtectR™ tubes can be used as a secondary shipping tube for multiple small volume tubes. With their airtight, leak-resistant secure seal, ProtectR™ tubes provide a protective barrier against CO2 exposure, safeguarding the sample(s) inside from costly damage. Only ProtectR™ tubes ensure consistent sample integrity and protein activity across experiments. Even if samples are delayed during shipping, they will not become compromised, saving time and money.