Learn More
Cole-Parmer Cole-Parmer Quartz Crucible, 50 mL, 1 ea
Supplier:  Cole-Parmer 3452634
Encompass_Preferred
Clear quartz glass crucibles are ideal for your applications that require ashing of samples!
Catalog No. NC1244109