Learn More
Chemglass Life Sciences Flask, Round Bottom, 50mL, Heavy Wall, 3-Neck, 19/22 CN - 19/22 SN's Angled 20°
SDP
Catalog No. CG1524A07
Encompass_Preferred
Flask, Round Bottom, 50mL, Heavy Wall, 19/22 - 19/22, 3-Neck, Angled 20¿.Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 19/22Side neck: 19/22
Catalog No. CG1524A07 Supplier Chemglass Life Sciences Supplier No. CG1524A07
Flask, Round Bottom, 50mL, Heavy Wall, 19/22 - 19/22, 3-Neck, Angled 20¿.Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 19/22Side neck: 19/22