Learn More
Chemglass Life Sciences Reaction Vessel Lid, 4-Nk, 50mL, Equivalent to Mettler Easymax Part # 51161032
SDP
Catalog No. 501532047
Encompass_Preferred
Reaction Vessel Lid, 4-Nk, 50mL, Compatible with Mettler Systems. (1) 14/20 SN, (1) GL-14 SN, (1) 10/18 SN and (1) 10/10 SN; Equivalent to Mettler Part # 51161032
Catalog No. 50-153-2047 Supplier Chemglass Life Sciences Supplier No. CGMR50L1
Reaction Vessel Lid, 4-Nk, 50mL, Compatible with Mettler Systems. (1) 14/20 SN, (1) GL-14 SN, (1) 10/18 SN and (1) 10/10 SN; Equivalent to Mettler Part # 51161032